Order Entry
Czech Republic
ContactUsLinkComponent
1,10-Phenanthroline monohydrate 99% for synthesis
Toxic
Environment
1,10-Phenanthroline monohydrate 99% for synthesis
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Environment
1,10-Phenanthroline monohydrate 99% for synthesis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

Related Information

Frequently Bought Together