ContactUsLinkComponent
Rotor package, Avanti JS-5.3
Rotor package, Avanti JS-5.3
Catalog # 521-4165
Supplier:  Beckman Coulter
undefined
Rotor package, Avanti JS-5.3
Catalog # 521-4165
Supplier:  Beckman Coulter
Specifications
  • Capacity:
    Nominal capacity
    2000 ml
  • Description:
    JS-5.3 swinging bucket (high performance) aluminium rotor package with microplate kit. For use in Avanti J-E and J-26 XP Series centrifuges only. Package includes rotor, buckets, tie-down knob, and microplate carrier kit (368914)
  • Pk:
    1
  • For:
    Four-place rotor with anodized aluminium buckets designed for use with adapters to accommodate a wide range of tubes and bottles. Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and blood sample preparation
  • Max. RPM:
    5300