ContactUsLinkComponent
Rotor package, Avanti J-Lite JLA-8.1000
Rotor package, Avanti J-Lite JLA-8.1000
Catalog # 521-4261
Supplier:  Beckman Coulter
undefined
Rotor package, Avanti J-Lite JLA-8.1000
Catalog # 521-4261
Supplier:  Beckman Coulter
Specifications
  • Description:
    J-LITE JLA-8.1000 rotor package. Includes rotor body, lid, carbon fibre canisters, labware kit (bottle rack, spatula, bottle grip and wrench 392574), and 6 polypropylene bottles with cap assemblies (3× C31597)
  • Pk:
    1
  • Max. RPM:
    8000
  • No. of tubes accepted:
    6×1000 ml/95×191 mm (3,8×7,65")