ContactUsLinkComponent
Rotors, Avanti JLA-16.250
Rotors, Avanti JLA-16.250
Supplier:  Beckman Coulter
Certificates
undefined
Rotors, Avanti JLA-16.250
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-4029
  • 521-4030
  • Capacity:
  • Nominal capacity
    1500 ml
  • Nominal capacity
    1500 ml
  • No. of tubes accepted:
  • 6×250 ml
  • Radius:
  • 134 mm
  • 134 mm
  • Max. RPM:
  • 16000
  • 16000