ContactUsLinkComponent
Rotor package, Avanti JA-30.50
Rotor package, Avanti JA-30.50
Supplier:  Beckman Coulter
Certificates
undefined
Rotor package, Avanti JA-30.50
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-3999
  • 521-4015
  • Capacity:
  • Nominal capacity
    400 ml
  • Nominal capacity
    400 ml
  • No. of tubes accepted:
  • 8×50 ml/29×104 mm (8×50 ml/1×4")
  • 8×50 ml/29×104 mm (8×50 ml/1×4")
  • Radius:
  • 108 mm
  • 108 mm
  • Max. RPM:
  • 30000
  • 30000