


2,4-Difluorobenzylmagnesium bromide 0.25 M in 2-MeTHF (2-methyltetrahydrofuran)
Catalog # H54966.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mL
Specifications
About this item
:
:231.323 g/mol
Storage Temperature:Ambient
UN:2924
ADR:"3",III
:GHS02
:GHS05
:GHS07
CAS Number:546122-71-8
Formula:C₇H₅BrF₂Mg
MDL Number:MFCD07698746
ADR:3,III