Order Entry
Czech Republic
ContactUsLinkComponent
LC-SMCC (N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]hexanoate) ≥90%, Sigma-Aldrich®
LC-SMCC (N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]hexanoate) ≥90%, Sigma-Aldrich®
Catalog # SIAL803383-50MG
Supplier:  Merck
CAS Number:  
LC-SMCC (N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]hexanoate) ≥90%, Sigma-Aldrich®
Catalog # SIAL803383-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :447.49 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:125559-00-4
Formula:C₂₂H₂₉N₃O₇
MDL Number:MFCD01863459