GMBS (N-Succinimidyl 4-maleimidobutyrate) ≥98% (by HPLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:80307-12-6
:
Formula:C₁₂H₁₂N₂O₆
MDL Number:MFCD00036817
:280.24 g/mol
Melting Pt:123…129 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
Frequently Bought Together