Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate) ≥90%, Sigma-Aldrich®
Catalog # SIAL803316-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:527.567 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:169751-10-4
Formula:C₁₈H₂₂N₃NaO₈S₃
MDL Number:MFCD00083162