Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Dimethylbarbituric acid ≥98.5%, GPR RECTAPUR®
Corrosive
Irritant
1,3-Dimethylbarbituric acid ≥98.5%, GPR RECTAPUR®
Catalog # 88368.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
1,3-Dimethylbarbituric acid ≥98.5%, GPR RECTAPUR®
Catalog # 88368.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    98,5 - 101,5 %
  • Assay (GC) (silylated):
    ≥ 98,5 %
  • Identification (FTIR):
    Conforms
  • Melting point:
    120,0 - 126,0 °C
  • Water:
    ≤ 6%

Specification Test Results

About this item

 :
 :156.14 g/mol
EINECS:212-211-7
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:769-42-6
Formula:C₆H₈N₂O₃
MDL Number:MFCD00006675