1,2-Distearoyl-sn-glycero-3-phosphocholine ≥99% (by TLC)
SIAL850365P-500MG
: Merck
:
- Pk:500 mg
- Seq:2725
- TLC, Purity:>99%
- TLC, Ninhydrin:Negative
- TLC, Iodine:One Major Spot
- TLC, Phosphorus:Positive
- TLC, Charring:Negative
- TLC, Water Dip:One Major Spot
- FAME (GC/FID), NLT 99%:18:0 (AUC)
:
:790.16 g/mol
:Freezer
:816-94-4
:C₄₄H₈₈NO₈P
:MFCD00036905