Voda, Molecular Biology Grade nuclease, DNase-, RNase-free, sterile
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
Specification Test Results
- Sterile (Bioburden test) :PASS
- DNAses free:NONE
- RNAses free:NONE
- Proteases free:NONE
- Human DNA free:NONE
Specification Test Results
Frequently Bought Together