Ammonium iron(III) sulphate dodecahydrate ACS
SIAL221260-500G
: Merck
:
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
:
:482.2 g/mol
:12,00549
:230 °C (1013 hPa)
:39…41 °C
:1.71 g/cm³ (20 °C)
:Ambient
:233-382-4
:GHS07
:7783-83-7
:NH₄Fe(SO₄)₂·12H₂O
:MFCD00150004