Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Selenium ≥99.5%

Selenium ≥99.5%

Poly bottle, 250 g
Catalog # SIAL209651-250G
Toxic
Health
Supplier:   Merck
Selenium ≥99.5%
Catalog # SIAL209651-250G
Supplier:  Merck
CAS Number:  7782-49-2
Selenium ≥99.5%

Poly bottle, 250 g

CAS Number:  7782-49-2
Price
6 550,00 Kč
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Boiling Pt:684.9 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.81 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • UN:3283
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    250 g

Specifications