ContactUsLinkComponent
Double coated tape, GPT-020F
Double coated tape, GPT-020F
  MMMAGPT-020F1540
 :  3M
undefined
Double coated tape, GPT-020F
  MMMAGPT-020F1540
 :  3M
  • Width:
    1540 mm
  • Colour:
    Transparent
  • Description:
    Double coated tape, GPT-020F
  • Length:
    50 m
  • Pk:
    1 Roll
Designed for versatility, 3M™ double coated tape GPT-020F is a transparent, 200 µm thick, modified solventless acrylic adhesive with a white printed PP filmic liner. It combines a high level of peel and shear performance, enabling high adhesion to a wide variety of substrates.
  • Providing the capability to bond to a wide variety of substrates, from high to low surface
  • High initial tack and humidity resistance
  • Easy handling and converting due to filmic liner delivery
  • 200 µm thick
This tape offers high initial tack and humidity resistance, providing easy handling and converting due to the filmic liner delivery. High temperature resistance: 190 °C short term and 90 °C long term.