Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IgG Mouse Monoclonal Antibody (DyLight® 800) [clone: eB182]
  ROCK18-4516-32
undefined
Anti-IgG Mouse Monoclonal Antibody (DyLight® 800) [clone: eB182]
  ROCK18-4516-32

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Monoclonal
  • Clone:
    eB182
  • Conjugation:
    DyLight® 800
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    Protein G purified
  • Pk:
    100 µl

 

 

Rabbit IgG TrueBlot® is a unique DyLight™ 800 conjugated anti-rabbit IgG immunoblotting (second step) reagent. Rabbit IgG TrueBlot® enables detection of immunoblotted target protein bands, without hindrance by interfering immunoprecipitating immunoglobulin heavy and light chains. It is easy to generate publication-quality IP/Fluorescent Western Blot data with Rabbit IgG TrueBlot®, simply substitute the conventional DL800 anti-rabbit IgG blotting reagent with Fluorescent Rabbit TrueBlot® Antibody DyLight™ 800 and follow the prescribed protocol for sample preparation and immunoblotting. Ideal for Li Cor Odyssey imaging as well as other IR and near IR imaging systems.

Rabbit IgG TrueBlot® is ideal for use in protocols involving immunoblotting of immunoprecipitated proteins. TrueBlot preferentially detects the non-reduced form of rabbit IgG over the reduced, SDS-denatured form of IgG. When the immunoprecipitate is fully reduced immediately prior to SDS-gel electrophoresis, reactivity of Rabbit IgG TrueBlot® with the 55 kDa heavy chains and the 23 kDa light chains of the immunoprecipitating antibody is minimised thereby eliminating interference by the heavy and light chains of the immunoprecipitating antibody in IP/Western blotting applications. Applications include studies examining post-translational modification (e.g., phosphorylation or acetylation) or protein-protein interactions. Fluorescent Rabbit TrueBlot® Antibody DyLight™ 800 may also be used for detection in immunoassays that do not employ immunoprecipitation.

Fluorescent Rabbit TrueBlot® Antibody DyLight™ 800 is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls). If used conservatively at 2.5mls/blot, cat. 18-4516-32 (100μL) will yield enough reagent for 200 blots.

Note that there are three key procedural considerations:
1. Protein A or G should not be used for the immunoprecipitation. For immunoprecipitation, anti-rat IgG beads, or anti-rabbit IgG beads should be used for rat or rabbit immunoprecipitating antibodies, respectively.
2. Immunoprecipitate should be completely reduced.
3. Bovine Serum Albumin, at low concentrations, should be used as the blocking protein for the immunoblot. DO NOT USE MILK.

Fluorescence technology is widely used to detect proteins. However, many common visible fluorophores often result in considerable background fluorescence in the visible range. Visible fluorophores are rarely used for membrane-based protein detection because of this high background. DyLight™ 800 and DyLight™ 680 antibody and reagent conjugates are specifically designed for protein detection methods that use longer-wavelength, near-infrared (IR) fluorophores to visualize proteins in western blotting and other applications. Very low background fluorescence in the IR range provides for a much higher signal-to-noise ratio than visible fluorophores. Detection levels in the picogram range on Western blots rival the sensitivity of chemiluminescence on film. DyLight™ 800 conjugates are optimized for the Odyssey® Infrared Imaging System developed by LI-COR. DyLight™ 800 conjugates are also suitable for immunofluorescence microscopy using commercially available excitation/emission filters in the 780nm/820nm range. Dual simultaneous labeling in western blots or microscopy is achieved when DyLight™ 800 conjugates are used in conjunction with DyLight™ 680 conjugates. DyLight™ 800 and DyLight™ 680 conjugates provide an ultra-sensitive and convenient alternative to standard chemiluminescent protein detection methods, as well as a valuable tool for multicolor imaging.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB182
Conjugation: DL800
Epitope:
Host: Mouse
Isotype:
Reactivity: Rabbit