Mouse IgG1 Isotype Control [clone: MOPC-21] (APC (Allophycocyanin)-Cy7®)
25-4714-U100
- Antibody type:Control
- Clone:MOPC-21
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8 °C; Protect from Light; Do Not Freeze.
- Tested applications:Flow Cytometry
- Pk:100 µG
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: MOPC21
Clonality:
Clone: MOPC-21
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: