Order Entry
Czech Republic
ContactUsLinkComponent
Hazard warning tape 766
Hazard warning tape 766
Catalog # MMMA766-YELL/BLACK
Supplier:  3M
undefined
Hazard warning tape 766
Catalog # MMMA766-YELL/BLACK
Supplier:  3M

Specifications

  • Width:
    50 mm
  • Colour:
    Yellow/black
  • Length:
    33 m
  • Pk:
    12 Roll

Specifications

About this item

3M™ Hazard Warning Tape 766 is a black and yellow striped vinyl tape. This tape is used for marking hazardous areas, protruding equipment, or preventing accidents in factories, warehouses, and hospitals.

  • Use for taping in all non-critical applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas

Identify and warn with Hazard Warning Tape 766. This distinctive yellow and black striped vinyl tape is coated with a rubber adhesive and designed for use as a marking tape for a variety of warning applications. The rubber adhesive offers quick stick and good holding strength on many substrates. Unlike messy paint solutions, this vinyl tape is quick to install and easy to clean-up. Conformability and instant adhesion makes this product a good choice for marking and warning applications on floors, equipment, walls, and pipes.

Recommended applications include floor marking, marking equipment pinch points, temporary floor marking around spills, and safety markings.

The tape features abrasion resistance to excel in light-duty identification of equipment and parts. These features, combined with easy removal properties, often makes this product a faster, more versatile, and less costly option than painting when used in marking and coding applications.