
3-Hydroxytyrosol ≥98% (by HPLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10597-60-1
:
Formula:C₈H₁₀O₃
MDL Number:MFCD01320529
:M-Clarity™ quality level = MQ100
:154.17 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together