

3,3'-Iminobis(N,N-dimethylpropylamine)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6711-48-4
:
Formula:C₁₀H₂₅N₃
MDL Number:MFCD00014880
:187.33 g/mol
Boiling Pt:128.0°C ...131.0°C (27.0 hPa)
Density:0.8410 g/mL
Flash Pt:98°C
EINECS:229-761-9
UN:2922
ADR:8,II
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to brown liquid
- Infrared spectrum:Conforms
- Refractive index:1.4470 to 1.4510 (20°C, 589 nm)
- Titration Total base:96.0 to 104.0 %
Specification Test Results
Frequently Bought Together