QuEChERS extraction salts, Q-sep®
Catalog # REST25849
Supplier: Restek
CAS Number:
Specifications
- Pk:50
- Description:Q-sep® QuEChERS extraction salt packets only (EN)
- Seq:03
- Size:50 packets
- Contents:4 g MgSO₄, 1 g NaCl, 1 g TSCD, 0,5 g DHS
Specifications
About this item
QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep® products make QuEChERS procedures even simpler.
- Free-flowing salts transfer easily and completely
- Easy-open packets eliminate the need for a second empty tube for salt transfer
- Convenient slim packets fit perfectly into tubes to prevent spills
- Ready-to-use tubes; no glassware required
- Pre-weighed, ultra-pure extraction salts
- Ideal for original unbuffered, AOAC (2007.01), and European (EN 15662) QuEChERS methods
No specialised glassware is required when using Q-sep® extraction packets and tubes. Free-flowing extraction salts and salt packets fit easily into the extraction tubes to make transferring salts to sample mess-free and easy.