Anti-EphB4 mouse monoclonal antibody [clone: 5B5]
Catalog # NOVUNBP1-47434
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:EPH Receptor B4
- Antigen symbol:EphB4
- Clonality:Monoclonal
- Clone:5B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:2050
- Antigen synonyms:soluble EPHB4 variant 1|soluble EPHB4 variant 2|EC 2.7.10|EC 2.7.10.1|hepatoma transmembrane kinase|Tyrosine-protein kinase TYRO11|MYK1|ephrin type-B receptor 4|HTKephrin receptor EphB4|Tyrosine-protein kinase receptor HTK|EPH receptor B4|Tyro11|EphB4|soluble EPHB4 variant 3
- Storage buffer:Ascites
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:Purified recombinant extracellular fragment of human EphB4 fused with hIgGFc tag expressed in HEK293 cell line.
- Purification:Unpurified
- Pk:0,1 mL
Specifications
About this item
The EphB4 Antibody (5B5) from Novus Biologicals is a mouse monoclonal antibody to EphB4. This antibody reacts with human. The EphB4 Antibody (5B5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: EphB4
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human