Order Entry
Czech Republic
ContactUsLinkComponent
Rat IgG2b Isotype Control [clone: LTF-2]
Rat IgG2b Isotype Control [clone: LTF-2]
Katalogové číslo 40-4031-U100
Supplier:  Tonbo Biosciences
undefined
Rat IgG2b Isotype Control [clone: LTF-2]
Katalogové číslo 40-4031-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Antigen symbol:
    LTF-2
  • Clone:
    LTF-2
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Neutralisation:
    Yes
  • Storage temperature:
    Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
    Functional Assays, Immunofluorescence Microscopy
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: LTF-2
Clonality:
Clone: LTF-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: