Order Entry
Czech Republic
ContactUsLinkComponent
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin)-Cy5®)
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin)-Cy5®)
Catalog # 55-4321-U100
Supplier:  Tonbo Biosciences
undefined
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin)-Cy5®)
Catalog # 55-4321-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Antigen name:
    UDP-glucuronosyltransferase 2A3
  • Clone:
    2A3
  • Conjugation:
    PE (Phycoerythrin)-Cy5®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Neutralisation:
    Yes
  • Storage temperature:
    Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
    Flow Cytometry
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.