Anti-HBsAg Rabbit polyclonal antibody
Catalog # NOVUNB100-62652
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis B Virus Surface Antigen (ayw)
- Antigen symbol:HBsAg
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Gene ID:944569
- Antigen synonyms:HBV major surface antigen|HBV surface antigen|protein S|HBsAg|Hepatitis B Virus major surface antigen|Hepatitis B Virus Surface Antigen|Major surface antigen
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:4,0 mg/ml
- Immunogen:Hepatitis B subtypes ad and ay.
- Purification:Protein A purified
- Pk:0,5 mL
Specifications
About this item
Rabbit Polyclonal HBsAg Antibody. Tested Applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Virus.
Type: Primary
Antigen: HBsAg
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus