Order Entry
Czech Republic
ContactUsLinkComponent
Anti-OTUD1 Rabbit Polyclonal Antibody
Anti-OTUD1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-90484
Supplier:  Novus Biologicals
Anti-OTUD1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-90484
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    OTUD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Gene ID:
    220213
  • Antigen synonyms:
    OTU domain containing 1|OTDC1
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:SSAAEPVIVSRSDPRDEKLALYLAEVEKQDKYLRQRNKYRFHIIPDGNCLYRAVSKTVYGDQSLHRELREQTVHYIADHLDHFSPLIEGDVGEFIIAAAQD
  • Purification:
    Immunogen affinity purified
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

The OTUD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OTUD1. This antibody reacts with human. The OTUD1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: OTUD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human