Anti-INPP5A Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-89361
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:INPP5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:3632
- Antigen synonyms:CTCL tumor antigen HD-CL-02,5PTASE|EC 3.1.3.56|InsP3 5-phosphatase|DKFZp434A1721|MGC116947,5PTase|MGC116949|type I inositol-14,5-trisphosphate 5-phophatase|inositol trisphosphate-5-phosphatase|43 kDa inositol polyphosphate 5-phophatase|inositol polyphosphate-5-phosphatase|40kD|type I inositol-14,5-trisphosphate 5-phosphatase|40kDa
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:EVVKLIFRESDNDRKVMLQLEKKLFDYFNQEVFRDNNGTALLEFDKELSVFKDRLYELDISFPPSYPYSEDARQGEQYMNTRCPAWCDRILMSPSAKELVLRSESEEKVVTYDHIGPNVCMGDHKPVFL
- Purification:Immunogen affinity purified
- Size:100 μl
- Pk:0,1 mL
Specifications
About this item
The INPP5A Antibody from Novus Biologicals is a rabbit polyclonal antibody to INPP5A. This antibody reacts with human. The INPP5A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: INPP5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human