Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-Aquaporin-7 Rabbit Polyclonal Antibody
Anti-Aquaporin-7 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-54384
Supplier:  Novus Biologicals
undefined
Anti-Aquaporin-7 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-54384
Supplier:  Novus Biologicals

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Aquaporin-7
  • Symbol antigenu:
    AQP7
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Identifikační číslo genu:
    364
  • Synonyma antigenu:
    Aquaporin-7-like|AQP7L|Aquaglyceroporin-7|AQP-7|aquaporin-7|AQP9MGC149556|aquaporin 7|Aquaporin adipose|AQPapaquaglyceroporin-7|MGC149555
  • Skladovací pufr:
    PBS & 2% Sucrose,
  • Skladovací teplota:
    Store at -20 °C. Avoid freeze-thaw cycles.
  • Imunogen:
    Synthetic peptides corresponding to AQP7 (aquaporin 7) The peptide sequence was selected from the C terminal of AQP7. Peptide sequence DSVAYEDHGITVLPKMGSHEPTISPLTPVSVSPANRSSVHPAPPLHESMA.
  • Čištění:
    Immunogen affinity purified
  • Rozměry protilátky:
    100 μl
  • Bal.:
    100 µl

Specifications

About this item

The Aquaporin-7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aquaporin-7. This antibody reacts with human. The Aquaporin-7 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Aquaporin-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human