ContactUsLinkComponent
VWR®, Sterile Masks
VWR®, Sterile Masks
Catalog # 76470-110
Supplier:  VWR Collection
VWR®, Sterile Masks
Catalog # 76470-110
Supplier:  VWR Collection
Specifications
  • Colour:
    White
  • Size:
    7W × 4H
  • Pk:
    200
Specifications
About this item
VWR® made in USA sterile masks are an ideal option for single-use needs.
  • Individually bagged
  • Available with ear loop or tie configurations
  • 3-ply construction
  • Gamma irradiated to 10⁻⁶
  • Enclosed plastic encapsulated nose wire
A key part of the VWR® made in USA PPE line, these sterile cleanroom masks provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
Packaging: Each mask individually bagged sealed. 10 bags per poly pack. 20 packs per case. Case is poly lined.