Silica gel, GPR RECTAPUR® 0,2 - 2,0 mm
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7631-86-9
:112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:> 1600 °C
Storage Temperature:Ambient
Specification Test Results
- Moisture loss :≤2,0%
- Adsorption capacity (H₂O; 40% RH):≥20,0%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC27616.292 spec_VWRC27616.360
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together