ContactUsLinkComponent
Perfect-it microfibre wipes, 2022 series, Scotch-Brite™
Perfect-it microfibre wipes, 2022 series, Scotch-Brite™
Catalog # MMMA2022
Supplier:  3M
undefined
Perfect-it microfibre wipes, 2022 series, Scotch-Brite™
Catalog # MMMA2022
Supplier:  3M
Specifications
  • Colour:
    Yellow
  • L×W:
    320×360 mm
  • Seq:
    02
  • Pk:
    100
Specifications
About this item
The 2022 series Scotch-Brite™ Perfect-it microfibre wipes are premium microfibre polishing cloths. They are ideal for cleaning and polishing a wide range of surfaces such as stainless steel, chrome, glass, enamel, plastic, and lacquered furniture.
  • Microfibre wipes ideal for cleaning and polishing any surface
  • Provides a high-quality, lint-free finish without streaks or marks
Clean and polish any surface with Scotch-Brite™ Perfect-it Microfibre Wipes, 2022 series. These premium microfibre polishing cloths are ideal for cleaning and polishing.