ContactUsLinkComponent
PrimeLock® High-Purity PFA Fittingss Accessories
PrimeLock® High-Purity PFA Fittingss Accessories
Certificates
PrimeLock® High-Purity PFA Fittingss Accessories
Specifications
  • Compression size / tube ext.Ø (port 1):
  • 1"
  • 1"
  • 1"
  • Connection type:
  • Flare Installation Insert
  • Flare
  • Flare Wrench
  • Max. operating pressure:
  • 120 psi at 70 °F (21 °C)
  • 120 psi at 70 °F (21 °C)
  • Max. temperature:
  • 200 °C (392 °F)
  • 200 °C (392 °F)
  • Min. temperature:
  • –18 °C (0 °F)
  • –18 °C (0 °F)
  • Warranty:
  • 2 months
  • 2 months
  • 2 months