ContactUsLinkComponent
Cadence Luer Adapter Fittings, Plated Brass
undefined
Cadence Luer Adapter Fittings, Plated Brass
  • Connection type (port 1):
  • Male luer slip
  • Male luer lock
  • Male luer slip
  • Male luer lock
  • Male luer slip
  • Male luer slip
  • Female luer
  • Male luer lock
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Male luer lock
  • Male luer lock
  • Female luer
  • Male luer lock
  • Female luer
  • Female luer
  • Male luer lock
  • Male luer lock
  • Female luer
  • Connection type (port 2):
  • 1/4" Barb
  • 1/4" NPT(M)
  • Male luer slip
  • 1/8" NPT(M)
  • Plug
  • 1/16" Barb
  • 1/4" NPT(M)
  • 1/8" Barb
  • 1/8" NPT(M)
  • 3/16" Barb
  • 1/8" Barb
  • Male luer lock
  • 1/4" Barb
  • Plug
  • 10-32 UNF
  • 10-32 UNF
  • Plug
  • Female luer
  • 1/4-28 UNF
  • Male luer lock
  • 1/4-28 UNF
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 1
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 1
  • 2
  • 2
  • 1
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 6 Months
  • 6 Months
  • 6 Months
  • 6 Months
Frequently Bought Together