ContactUsLinkComponent
High Volume Chemical Resistant Siphon Pump, 5 strokes/gallon, 2" IBS adapter
High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
Catalog # MFLX06433-72
undefined
High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
Catalog # MFLX06433-72
Specifications
  • Flow rate:
    5 strokes/gal.
  • Description:
    High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
  • Wetted materials:
    Solvent-welded PVC body and shaft;vinyl hose; Viton valves and seals
  • Tube length, inlet:
    38"
  • Warranty:
    1 Month
  • Pk:
    1
Specifications
About this item
Ideal for most acids, bases, alcohols, aldehydes and aliphatic hydrocarbons.
  • PVC with Viton® valves and seals for chemical resistance
  • Withstands temperatures upto 120F (49C)
  • Self-priming
This hand-operated drum pump is ideal for pumping most acids, bases, alcohols, aldehydes and aliphatic hydrocarbons. Constructed of PVC with Viton® valves and seals for optimal chemical resistance, this pump offers 5 strokes/gallon.
  • Flow rate:
    5 strokes/gal.
  • Description:
    High Volume Chemical Resistant Siphon Pump, 5 strokes per gallon, 2" IPS adapter
  • Environmentally Preferable:
  • Wetted materials:
    Solvent-welded PVC body and shaft;vinyl hose; Viton valves and seals
  • Tube length, inlet:
    38"
  • Warranty:
    1 Month
  • Pk:
    1