ContactUsLinkComponent
Noshok® Metal Mini Needle Valves
Noshok® Metal Mini Needle Valves
Certificates
undefined
Noshok® Metal Mini Needle Valves
Specifications
  • Orifice:
  • 4,3688 mm (0,172")
  • 4,3688 mm (0,172")
  • 4,3688 mm (0,172")
  • 4,3688 mm (0,172")
  • 4,3688 mm (0,172")
  • Wetted materials:
  • Brass
  • 316 Stainless Steel
  • Brass
  • Brass
  • Brass
  • Working pressure:
  • 6000 psi
  • 10000 psi
  • 6000 psi
  • 3000 psi
  • 6000 psi
  • Warranty:
  • 3 years
  • 3 years
  • 3 years
  • 3 years
  • 3 years