Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Power:120 VAC; 60 Hz
- Max. viscosity:1200 cP
- Description:Drum pump motor, TEFC, 2/3 hp, 120 VAC, 60 Hz
- CSA regulatory approval:Yes
- Motor hp:2/3
- UL Canada regulatory approval:Yes
- UL US regulatory approval:Yes
- Motor type:TEFC
- Warranty:1 year
- Pk:1
Specifications
About this item
Mix and pump emulsions and suspensions in a single operation.
- For thin-bodied and low-viscosity liquids
- Small pump tube diameter is ideal for use in applications with narrow-necked containers
- Ensures media are properly mixed and homogenized
Mixing intensity and delivery rate of the pump are determined by the rate of flow, viscosity, and density of the fluid. Drive shafts of the pump tubes are sealed by a single-acting mechanical seal and the two shaft seals behind it. Pump tubes are non-lubricated to prevent any contamination of fluids. Model 70512-10 is a sealless pump tube design.
Interchange multiple pump tubes with one motor. For hazardous-duty applications, use pump tube 70512-20 with pump motors 70513-30, -40, -50, or -60.
Note: Do not allow pump tubes to run dry.
- Power:120 VAC; 60 Hz
- Max. viscosity:1200 cP
- Description:Drum pump motor, TEFC, 2/3 hp, 120 VAC, 60 Hz
- Environmentally Preferable:
- CSA regulatory approval:Yes
- Motor hp:2/3
- UL Canada regulatory approval:Yes
- UL US regulatory approval:Yes
- Motor type:TEFC
- Warranty:1 year
- Pk:1