ContactUsLinkComponent
Dual and Quad-Head Diaphragm Pumps
Dual and Quad-Head Diaphragm Pumps
Certificates
undefined
Dual and Quad-Head Diaphragm Pumps
Specifications
  • Duty cycle continuous:
  • Yes
  • Yes
  • Inlet connection:
  • 1/4" hose barb
  • 1/4" hose barb
  • Max. temperature:
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • Motor type:
  • Brushless DC
  • Brushless DC
  • Outlet connection:
  • 1/4" hose barb
  • 1/4" hose barb
  • Power pigtail connection:
  • Yes
  • Yes
  • Run dry:
  • Yes
  • Yes
  • Self-priming:
  • Yes
  • Yes
  • Wetted materials:
  • Polypropylene, Silicone
  • Polypropylene, Silicone
  • Warranty:
  • 1 year
  • 1 year