Sanitary Tri-Clamp End Caps, Stainless Steel, 3/4"
MFLX30508-11
6 month warranty
- Connection Type:Sanitary Cap
- Flow pattern:Straight
- Sanitary clamp size (Port 1):3/4"
- Description:Sanitary Tri-Clamp End Caps, Stainless Steel, 3/4"
- Warranty:6 Months
- Pk:1
- Fitting material:Stainless Steel 316
- Sanitary gasket size:3/4"
Sanitary seals open end to prevent debris from accumulating.
- Ouick and easy to mount or dismount with a pressure clamp
The tri-clamp end caps play an important role in the sanitation process, allowing an open end to be sanitary seals preventing additional debris from accumulating.
- Connection Type:Sanitary Cap
- Flow pattern:Straight
- Sanitary clamp size (Port 1):3/4"
- Description:Sanitary Tri-Clamp End Caps, Stainless Steel, 3/4"
- Environmentally Preferable:
- Warranty:6 Months
- Pk:1
- Fitting material:Stainless Steel 316
- Sanitary gasket size:3/4"