ContactUsLinkComponent
Pump Motor Spider Inserts, Buna-N®
Spider Insert, Buna-N, for A/L090/L095 Couplings
Catalog # MFLX07127-69
undefined
Spider Insert, Buna-N, for A/L090/L095 Couplings
Catalog # MFLX07127-69
Specifications
  • Description:
    Spider Insert, Buna-N, for A/L090/L095 Couplings
  • Pk:
    1
Specifications
About this item
Create your own motor mounting with the hardware below.
  • Constructed of durable Buna-N®
Spider smooths power transfer, absorbs vibration, and compensates for minor misalignment.
  • Description:
    Spider Insert, Buna-N, for A/L090/L095 Couplings
  • Environmentally Preferable:
  • Pk:
    1