Order Entry
Czech Republic
ContactUsLinkComponent
Pump Motor Coupling Hardware and Accessories
Coupling hardware for 5/8" diameter bore, 7.2 hp maximum
Katalogové číslo MFLX07127-59
undefined
Coupling hardware for 5/8" diameter bore, 7.2 hp maximum
Katalogové číslo MFLX07127-59

Specifications

  • Description:
    Coupling hardware for 5/8" diameter bore, 7,2 hp maximum
  • Pk:
    1
  • Motor hp:
    7,2

Specifications

About this item

Create your own motor mounting with the hardware below.

  • Optimize performance of your Pump Motor

Order one coupling with the same diameter as the motor driving shaft, one coupling with the same diameter as the driven shaft, and one spider (see accessories).

  • Description:
    Coupling hardware for 5/8" diameter bore, 7,2 hp maximum
  • Environmentally Preferable:
  • Pk:
    1
  • Motor hp:
    7,2