Order Entry
Czech Republic
ContactUsLinkComponent
Masterflex® L/S® Hazardous-Duty Fixed-Speed Drives, Avantor®
Masterflex® L/S® Hazardous-Duty Fixed-Speed Drive, 470 rpm; 115 VAC
Catalog # MFLX07571-30
2 year warranty
undefined
Masterflex® L/S® Hazardous-Duty Fixed-Speed Drive, 470 rpm; 115 VAC
Catalog # MFLX07571-30
2 year warranty
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Maximum Flow Rate:
    2700 ml/min
  • Power:
    115 VAC; 60 Hz
  • Control type:
    Fixed speed
  • Pump series:
    L/S
  • Description:
    L/S fixed-speed hazardous-duty drive; 470 rpm.
  • Drive height:
    232 mm (9,125")
  • Drive IP rating:
    IP 55
  • Drive length:
    406 mm (16")
  • Drive width:
    216 mm (8,5")
  • Motor hp:
    1/2
  • Speed control:
    ±1%
  • Type of pump heads accepted:
    Standard, Easy-Load, Easy-Load II, Quick Load, High Performance, Cartridge, Multichannel
  • Washdown capable:
    Yes
  • Motor:
    370 W
  • Speed:
    Fixed
  • Warranty:
    1 year
  • Pk:
    1
  • Max. RPM:
    470 rpm
  • No. of pump heads accepted:
    2

Specifications

About this item

Make hazardous-duty applications easy.

  • Use where special electrical precautions are necessary to ensure safe operation
  • Variable-speed drive provides a wide flow range for maximum flexibility
  • Motor is UL-listed for Class 1, Group D; Class 2, Groups F/G; and Division 1 hazardous locations—use where flammable gases/vapors may be present
  • Constructed with painted steel and aluminum housing for IP55-rating—protects against dust/low-pressure water jets

Fixed-speed hazardous-duty drives are an economical alternative to the variable-speed hazardous-duty drives. Their 1/2-hp motors make these drives ideal for multiple head applications—stack up to 2 pump heads for multichannel pumping. Drive accepts nine pump head types: Standard, Easy-Load®, Easy-Load® II, Easy-Load® 3, Multichannel, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm.

Delivery: Includes drive only. Power switch/line cord are not included. Safely connect power with your own protected connections.
  • Maximum Flow Rate:
    2700 ml/min
  • Power:
    115 VAC; 60 Hz
  • Control type:
    Fixed speed
  • Pump series:
    L/S
  • Description:
    L/S fixed-speed hazardous-duty drive; 470 rpm.
  • Environmentally Preferable:
  • Drive height:
    232 mm (9,125")
  • Drive IP rating:
    IP 55
  • Drive length:
    406 mm (16")
  • Drive width:
    216 mm (8,5")
  • Motor hp:
    1/2
  • Speed control:
    ±1%
  • Type of pump heads accepted:
    Standard, Easy-Load, Easy-Load II, Quick Load, High Performance, Cartridge, Multichannel
  • Washdown capable:
    Yes
  • Motor:
    370 W
  • Speed:
    Fixed
  • Warranty:
    1 year
  • Pk:
    1
  • Max. RPM:
    470 rpm
  • No. of pump heads accepted:
    2