Evsco Valves Manual and Actuated Accessories
Supplier: Avantor Fluid Handling
Certificates
About this item
Safely mount your plastic elliptical valves.
- Optimize performance of your Evsco Valves
Constucted of durable polypropylene, these mounting brackets are ideal for making your valve more versitile. These mounting brackets allow for wall, panel, or undermount.
- Environmentally Preferable:
Specifications