ContactUsLinkComponent
Biosafety containment cabinet, for CytoFLEX SRT, SterilGARD
Biosafety containment cabinet, for CytoFLEX SRT, SterilGARD
  BECMC71890
Biosafety containment cabinet, for CytoFLEX SRT, SterilGARD
  BECMC71890
  • Description:
    Biosafety containment cabinet, SterilGARD for CytoFLEX SRT, 230V
  • Pk:
    1
Class II Type biosafety containment cabinet designed specifically for the CytoFLEX SRT in collaboration with Baker Corporation, including aerosol evacuation functionality.
  • Personnel protection or containment by an intake air velocity of no less than 100 feet per minute (fpm) through the front access opening
  • Product protection by the HEPA (high efficiency particulate air) filtered downflow air inside the cabinet work area
  • Environmental protection by allowing only HEPA filtered to be exhausted to the room
For indoor use only.
 : CE, RUO, EN 12469, BS EN 12469, SNS 12469, NF-095, YY-0569, JIS K 3800, AS 1807.1:2009.