Order Entry
Czech Republic
ContactUsLinkComponent
 
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
  MDTC46-102-RF
 :  Corning
 :  
undefined
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
  MDTC46-102-RF
 :  Corning
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :238.31 g/mol
 :402 °C (1013 hPa)
 :109.5…110.5 °C
 :Freezer
 :367-93-1
 :C₉H₁₈O₅S
 :MFCD00063273