Order Entry
Czech Republic
ContactUsLinkComponent
 
Gentamicin sulphate solution 50 mg/ml in deionised water
Catalog # MDTC30-005-CR
Supplier:  Corning
CAS Number:  
undefined
Gentamicin sulphate solution 50 mg/ml in deionised water
Catalog # MDTC30-005-CR
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    10
  • Seq:
    0010

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:1405-41-0
MDL Number:MFCD00270181