Clip for tubes and cables, b.safe
For easy bundling and fixing of signal cables, grounding cables, waste tubes, and vent tubes inside of safety cabinets and built-in installations.
- Chemical resistance: ++ very good
- Material: POM
- Temperature resistance: from −30 to +100 °C
Clip made of mechanically tight POM for safe clipping of the different cables or tubes. Design for wall mount includes fastening material.
Frequently Bought Together