ContactUsLinkComponent
Mount for waste pipes, b.safe
  BOHLR1163-32
 :  Bohlender
Mount for waste pipes, b.safe
  BOHLR1163-32
 :  Bohlender
  • Description:
    b.safe mount for waste pipes
  • Pk:
    1
Set consisting of (1) base plate with female thread M8, (1) threaded rod M8, length 200 mm, (1) pipe clamp with female thread M8 for tube ext.Ø 32 mm, and fastening material for assembly on walls or under desktops.
  • Base plate dimensions: 80×30×3 mm
Fix the base plate on the wall. First connect the threaded rod with the pipe clamp, then connect the pipe clamp to the base plate. Make sure to align the clamp correctly. Open the clamp, clip in the b.safe Waste Pipe and close the clamp again.