ContactUsLinkComponent
CultiControl freeze-dried microorganism strains derived from NCTC® cultures
  70121.
 :  
CultiControl freeze-dried microorganism strains derived from NCTC® cultures
  70121.
 :  
  • Pk:
    5
  • Description:
    Serratia marcescens derived from NCTC 13382
  • Seq:
    89
Lyophilised reference stock culture preparations containing a single strain of a microorganism for quality control of culture media, education and proficiency programs. Lyophilisation is a method for long-term preservation of microorganisms. The microorganism preparations are derived from NCTC® (National Collection of Type Cultures by the UK Health Security Agency - UKHSA).
  • One single passage from the reference NCTC® culture
  • Two years shelf life
  • More than 100 freeze-dried microorganisms available
 : CultiControl microorganisms are packaged in a resealable vial that contains five (5) lyophilised pellets of a single microorganism strain, and a desiccant to prevent adverse accumulations of moisture.
 : These products are for in-vitro use only.