Order Entry
Czech Republic
ContactUsLinkComponent
VWR®, Penicillin-streptomycin solution (100X)
VWR®, Penicillin-streptomycin solution (100X)
Catalog # 392-0406
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Penicillin-streptomycin solution (100X)
Catalog # 392-0406
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    100 mL
  • Description:
    Penicillin-Streptomycin solution (100X)
  • Sterility:
    Sterile

Specifications

About this item

VWR® penicillin-streptomycin solution (100X) is a large spectrum antibiotic designed to control and destroy gram-negative and gram-positive bacteria.

  • With 10000 units penicillin and 10 mg/ml streptomycin in 0,9% NaCl
  • Sterile: 0,1 μm filtered
  • Use in cell culture applications at 10 ml/L for tissue culture media containing serum
  • Endotoxin level at 10 ml/L of solution: <0,1 EU/ml

Antibiotics, combined with good sterile technique of manipulation of cell culture, help prevent microbiological contamination.