Order Entry
Czech Republic
ContactUsLinkComponent
Piperidine ≥99.5% (GC, on anhydrous basis) for peptide synthesis
Piperidine ≥99.5% (GC, on anhydrous basis) for peptide synthesis
Supplier:  VWR Chemicals
Certificates
undefined
Piperidine ≥99.5% (GC, on anhydrous basis) for peptide synthesis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC on anhydrous basis):
    min. 99.5%
  • Color (APHA Pt-Co):
    max. 20
  • Pyridine:
    max. 0.15%
  • Residue after evaporation:
    max. 0.001% (w/w)
  • Water (Karl Fischer):
    max. 0.1% (w/w)
  • Iron (Fe):
    max. 0.1 ppm
  • Magnesium (Mg):
    max. 0.1 ppm
  • Lead (Pb):
    max. 0.1 ppm
  • Zinc (Zn):
    max. 0.1 ppm

Specification Test Results

Related Information

Frequently Bought Together